Butyl acetate ≥99.5%, liquid ACS

Supplier: Thermo Scientific Chemicals
Warning

046186-AP 046186-K2 046186-K7
AA46186-APEA 77.77 USD
AA46186-AP AA46186-K2 AA46186-K7
Butyl acetate ≥99.5%, liquid ACS
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
Boiling Pt: 124…126 °C
Melting Pt: –78 °C
Density: 0.881
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
Merck Index: 14,1535
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR