Tungsten ≥99.95%, rods, Diameter 2 mm (0.08 in)
Supplier: Thermo Scientific Chemicals
043971-BV
043971-CE
043971-9X
AA43971-BVEA
74.5
USD
AA43971-BV
AA43971-CE
AA43971-9X
AA43971-DAT
Tungsten ≥99.95%, rods, Diameter 2 mm (0.08 in)
Tungsten
~6.1g/10cm
Formula:
W MW: 183.84 g/mol Boiling Pt: 5660 Melting Pt: 3410 Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...