TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TAPSO, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid

H66236-14 H66236-22
AAH66236-14EA 28.9 USD
AAH66236-14 AAH66236-22
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
TAPSO (2-hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula: C₇H₁₇NO₇S
MW: 259.28 g/mol
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00038352
CAS Number: 68399-81-5
EINECS: 269-993-8
Merck Index: 15,9191

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR