Celite® 535, filter aid
Supplier: Thermo Scientific Chemicals
|
Warning
|
H37758-0B
H37758-0I
AAH37758-0BEA
97.59
USD
AAH37758-0B
AAH37758-0I
Celite® 535, filter aid
Celite® 535, filter aid
Formula:
SiO₂ Melting Pt: Density: 2.4 Storage Temperature: Ambient |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 EINECS: 272-489-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...