4-Isopropylbenzyl alcohol ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Isopropylphenyl)methanol

H36692-14 H36692-22
AAH36692-14EA 32.77 USD
AAH36692-14 AAH36692-22
4-Isopropylbenzyl alcohol ≥97%
4-Isopropylbenzyl alcohol
Formula: C₁₀H₁₄O
Boiling Pt: 135-136°C/26mm
Density: 0.980
Flash Pt: 110°C(230°F)
Storage Temperature: Ambient
MDL Number: MFCD00004663
CAS Number: 536-60-7
EINECS: 208-640-4
Merck Index: 14,2619

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR