Phenylphosphonic dichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide

H36037-14 H36037-22
AAH36037-14EA 42.62 USD
AAH36037-14 AAH36037-22
Phenylphosphonic dichloride 99%
Phenylphosphonic dichloride
Formula: C₆H₅Cl₂OP
MW: 194.98 g/mol
Boiling Pt: 258°C
Melting Pt: 1-3°C
Density: 1.379
Flash Pt: 204°(399°F)
Storage Temperature: Ambient
MDL Number: MFCD00002070
CAS Number: 824-72-6
EINECS: 212-534-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR