Chromium (III) sulphate hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-2414-250G 93-2414-50G
100206-976EA 213.72 USD
100206-976 100208-530
Chromium (III) sulphate hydrate
Chromium (III) sulphate hydrate
Formula: Cr₂O₁₂S₃·H₂O
MW: 392.17 g/mol (anhy)
MDL Number: MFCD00150215
CAS Number: 15244-38-9

Order Now

Specification Test Results

Form Powder
color Greenish-black


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR