Di-tert-butylphosphine ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
15-1040-5G
15-1040-25G
100204-942EA
239.2
USD
100204-942
100204-940
Di-tert-butylphosphine ≥98%
Di-tert-butylphosphine
Phosphine
Formula:
C₈H₁₉P Boiling Pt: 38…40 °C Melting Pt: –17 °C Density: 0.790 Flash Pt: –1 °F |
MDL Number:
MFCD00134241 CAS Number: 819-19-2 UN: 2845 ADR: 4.2,I |
Specification Test Results
Assay | 98+% |
Color | Colorless |
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...