Tergitol™ 15-S-9, Surfactant, Spectrum chemical

Supplier: Spectrum Chemicals
T1261-100ML T1261-4LTGL T1261-500ML T1261-20LT
700011-110EA 80.9 USD
700011-110 700011-116 700011-118 700011-114
Tergitol™ 15-S-9, Surfactant, Spectrum chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ 15-S-9, Surfactant is a nonionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier & detergent, enhances paint thickener efficiency and aids in rinse-ability of solvent-based systems. This product is suggested for use in cleaners, prewash spotters and paints & coatings.

  • Superior detergency
  • Soluble in water
  • Rapid dissolution & good rinseability
  • Low odor
  • Narrow gel range
  • Excellent formulation & handling properties
  • Readily biodegradable (OECD 301E)

Density at 20°C (68°F): 1.006 g/ml
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR