Metalguard® Medium Weight 1878 Cut Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
1878L 1878M 1878S 1878XL
80091-396EA 22.75 USD
80091-396 80091-398 80091-400 80091-402
Metalguard® Medium Weight 1878 Cut Resistant Gloves, Wells Lamont
Gloves
MetalGuard® medium weight gloves form to hands for comfortability.
  • Continuous knit glove forms to the hand
MetalGuard® medium weight gloves allow full range of movement, making them flexible and comfortable. Gloves are continuous, and they can be used as a liner or as a glove alone. Gloves are prewashed and preshrunk for a comfortable fit.
Certificaciones: ANSI Cut Level 4, A5.
Información para pedidos: These gloves are sold by the dozen pair.
Precaución: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.