Carbomer 940 resin NF

Supplier: Spectrum Chemical Mfg. Corp.

C1477-500GM C1477-2.5KG C1477-45KGBL C1477-100GM
95032-408EA 585.18 USD
95032-408 95032-406 700012-338 95032-404
Carbomer 940 resin NF
Carbomer 940

Carbomer 940, NF is a polyvinyl carboxy polymer. 


  • Synonyms: Carbopol 940


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Certificaciones: Packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD01941675
CAS Number: 9007-17-4

Order Now

Specification Test Results

Carboxylic acid groups (dried basis) 56.0 - 68.0%
Viscosity of a 0.5% solution 40,000 - 60,000 mPa
Loss on drying 2.00%
Heavy metals 20 µg/g
Benzene 0.50%
Identification Passes test
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR