Dodecenylsuccinic acid detergent

Supplier: BIOSYNTH INTERNATIONAL INC MS

Q-201034-1KG DIS-CONTINUED-25KG
102969-008EA 241.67 USD
102969-008 102969-010
Dodecenylsuccinic acid detergent
Dodecenylsuccinic acid

Used as biological detergents for laboratory use only not for food or drug use.

Formula: C₁₆H₂₈O₄
MW: 284.396 g/mol
CAS Number: 29658-97-7

Order Now

Specification Test Results

1H-NMR Spectrum corresponds to assigned structure
13C-NMR Spectrum corresponds to assigned structure
Purity (TLC) min. 97 area-%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR