LaboPlast®/SteriPlast® Micro Spatula, Bürkle
Supplier: BURKLE INC
5378-0032 5378-1032
75876-078PK 48.66 USD
75876-078 75876-080
LaboPlast®/SteriPlast® Micro Spatula, Bürkle
Spatulas
The Micro Spatula is a double scoop spatula with rounded ends, content 1.0 (0.03 oz.) and 0.25 ml (0.01 oz.)
  • Polystyrene construction
  • Production, assembly, and packaging according to Cleanroom Class 7
  • Complies with EU food requirements and FDA regulations
  • Optionally packaged individually and sterilized by gamma rays
The Micro Spatula can be used to sample, transfer, or process small amounts of chemicals, powders, granulates, pastes, creams, or liquids. A sturdy and convenient center handle facilitates precise work.
Envase: Cleanroom manufactured, Micro Spatulas are available in two packaging options. LaboPlast® Micro Spatulas are supplied in a large package with 100 pieces. SteriPlast® Micro Spatulas feature 100 pieces packaged individually and sterilized by gamma rays.