Wash Solution Concentrate, 20X, SeraCare
Supplier: KPL
5150-0008 51-50-0011
95059-132EA 255.04 USD
95059-132 95059-134
Wash Solution Concentrate, 20X, SeraCare
Buffers Electrophoresis Buffers Washing Buffers
Convenient 20X concentrate containing imidazole-buffered saline and Tween® 20
  • Suitable for ELISA, blotting, and immunohistochemistry applications
This is used a general immunoassay wash buffer.