Methyl-2-methyltetrahydrofuran-2-carboxylate

Supplier: Matrix Scientific

113223-1G 113223-5G
102891-692EA 436.92 USD
102891-692 102891-694
Methyl-2-methyltetrahydrofuran-2-carboxylate
Methyl-2-methyltetrahydrofuran-2-carboxylate

Hazard: Irritant

Formula: C₇H₁₂O₃
MDL Number: MFCD18651757
CAS Number: 1218915-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR