(3,4)-cis-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115876-500MG 115876-1G 115876-5G
102898-748EA 1318.46 USD
102898-748 102898-750 102898-752
(3,4)-cis-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate ≥95%
(3,4)-cis-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇FN₄O₂
MDL Number: MFCD22628732
CAS Number: 1070896-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR