3-(4-Iodophenyl)oxetan-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
115903-250MG
115903-500MG
115903-1G
102898-712EA
2267.69
USD
102898-712
102898-714
102898-716
3-(4-Iodophenyl)oxetan-3-amine hydrochloride ≥95%
3-(4-Iodophenyl)oxetan-3-amine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁ClINO |
MDL Number:
MFCD22628742 CAS Number: 1416323-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...