3-(4-Chlorophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115875-100MG 115875-250MG 115875-500MG
102898-544EA 1373.85 USD
102898-544 102898-744 102898-746
3-(4-Chlorophenyl)oxetane-3-carboxylic acid ≥95%
3-(4-Chlorophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉ClO₃
MDL Number: MFCD22544052
CAS Number: 1393534-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR