(R)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate ≥95%

Supplier: Matrix Scientific

115835-500MG 115835-1G 115835-5G
102898-498EA 1238.46 USD
102898-498 102898-500 102898-502
(R)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate ≥95%
(R)-Benzyl-1-(oxetan-3-yl)pyrrolidin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₃
MDL Number: MFCD21602104
CAS Number: 1349807-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR