Oxo-(1-triisopropylsilyl-1H-pyrrol-3-yl)acetic acid ethyl ester ≥95%

Supplier: Matrix Scientific

115799-1G 115799-5G 115799-25G
102898-134EA 140 USD
102898-134 102898-136 102898-138
Oxo-(1-triisopropylsilyl-1H-pyrrol-3-yl)acetic acid ethyl ester ≥95%
Oxo-(1-triisopropylsilyl-1H-pyrrol-3-yl)acetic acid ethyl ester

Hazard: Irritant

Formula: C₁₇H₂₉NO₃Si
MDL Number: MFCD22573394
CAS Number: 87630-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR