2,4-Dichloropyrimidine-5-carbaldehyde

Supplier: Matrix Scientific

113067-1G 113067-5G
102897-668EA 727.69 USD
102897-668 102897-670
2,4-Dichloropyrimidine-5-carbaldehyde
2,4-Dichloropyrimidine-5-carbaldehyde

Hazard: Irritant

Formula: C₅H₂Cl₂N₂O
MDL Number: MFCD11112096
CAS Number: 871254-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR