4,4-Difluoropiperidine-1-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

115770-250MG 115770-500MG 115770-1G
102897-482EA 560 USD
102897-482 102897-484 102899-652
4,4-Difluoropiperidine-1-sulfonyl chloride ≥95%
4,4-Difluoropiperidine-1-sulfonyl chloride

Hazard: Corrosive/Refrigerate

Formula: C₅H₈ClF₂NO₂S
MW: 219.64 g/mol
Boiling Pt: 74 °C (0.31 Torr)
MDL Number: MFCD18917106
CAS Number: 1224929-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR