6-(Aminomethyl)-2-methylthio-1H-pyrimidin-4-one ≥96%

Supplier: Matrix Scientific

115754-500MG 115754-1G 115754-5G
102897-396EA 495.38 USD
102897-396 102897-398 102897-400
6-(Aminomethyl)-2-methylthio-1H-pyrimidin-4-one ≥96%
6-(Aminomethyl)-2-methylthio-1H-pyrimidin-4-one

Hazard: Irritant

Formula: C₆H₉N₃OS
MDL Number: MFCD09754187
CAS Number: 1023812-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR