2-Azatricyclo[3.3.1.13,7]decane-5-carboxylic acid methyl ester hydrochloride ≥97%

Supplier: Matrix Scientific

115753-250MG 115753-500MG 115753-1G
102897-390EA 4807.69 USD
102897-390 102897-392 102897-394
2-Azatricyclo[3.3.1.13,7]decane-5-carboxylic acid methyl ester hydrochloride ≥97%
2-Azatricyclo[3.3.1.13,7]decane-5-carboxylic acid methyl ester hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClNO₂
MDL Number: MFCD19707044
CAS Number: 1389264-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR