3-Azabicyclo[3.1.1]heptan-6-ol hydrochloride ≥95%
Supplier: Matrix Scientific
115752-250MG
115752-500MG
115752-1G
102897-384EA
1538.46
USD
102897-384
102897-386
102897-388
3-Azabicyclo[3.1.1]heptan-6-ol hydrochloride ≥95%
3-Azabicyclo[3.1.1]heptan-6-ol hydrochloride
Hazard: Irritant
Formula:
C₆H₁₂ClNO |
MDL Number:
MFCD22415249 CAS Number: 1389264-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...