4-(Boc-aminomethyl)-6-chloropyrimidine ≥95%

Supplier: Matrix Scientific

115746-250MG 115746-500MG 115746-1G
102897-354EA 1512.31 USD
102897-354 102897-356 102897-358
4-(Boc-aminomethyl)-6-chloropyrimidine ≥95%
4-(Boc-aminomethyl)-6-chloropyrimidine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O₂
MW: 243.69 g/mol
CAS Number: 1404373-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR