2-Methyl-4-penten-2-ol ≥97%

Supplier: Matrix Scientific

115737-1G 115737-5G 115737-25G
102897-284EA 61.54 USD
102897-284 102897-286 102897-288
2-Methyl-4-penten-2-ol ≥97%
2-Methyl-4-penten-2-ol

Hazard: Irritant

Formula: C₆H₁₂O
MDL Number: MFCD00021813
CAS Number: 624-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR