3-Fluoro-3-oxetanemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-3-hydroxymethyloxetane, (3-Fluorooxetan-3-yl)methanol

115736-100MG 115736-250MG 115736-500MG
102897-278EA 192.31 USD
102897-278 102897-280 102897-282
3-Fluoro-3-oxetanemethanol ≥95%
3-Fluoro-3-oxetanemethanol

Hazard: Irritant

Formula: C₄H₇FO₂
MW: 106.1 g/mol
MDL Number: MFCD14706188
CAS Number: 865451-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR