6-Amino-2-azaspiro[3.3]heptane-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

115576-1G 115576-500MG
102896-868EA 5893.85 USD
102896-868 102899-500
6-Amino-2-azaspiro[3.3]heptane-6-carboxylic acid ≥95%
6-Amino-2-azaspiro[3.3]heptane-6-carboxylic acid

Hazard: Irritant

Formula: C₇H₁₂N₂O₂
MDL Number: MFCD16153811
CAS Number: 1170775-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR