rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3,4-trans-4-(benzylamino)-3-fluoropiperidine-1-carboxylate racemate

115473-250MG 115473-500MG
102896-428EA 1593.85 USD
102896-428 102896-430
rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine ≥95%
rel-(3R,4R)-4-Benzylamino-1-[tert-butoxycarbonyl]-3-fluoro piperidine

Hazard: Irritant

Formula: C₁₇H₂₅FN₂O₂
MW: 308.4 g/mol
MDL Number: MFCD20261376
CAS Number: 211108-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR