Benzyl-2-amino-7-azaspiro[3.5]nonane-7-carboxylate ≥95%

Supplier: Matrix Scientific

115472-500MG 115472-1G
102896-424EA 2143.08 USD
102896-424 102896-426
Benzyl-2-amino-7-azaspiro[3.5]nonane-7-carboxylate ≥95%
Benzyl-2-amino-7-azaspiro[3.5]nonane-7-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₂
MDL Number: MFCD15071709
CAS Number: 147611-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR