(1R,2S,5S)-Rel-3-azabicyclo[3.1.0]hexane-2-methanol ≥95%
Supplier: Matrix Scientific
115470-250MG
115470-500MG
115470-1G
102896-414EA
1153.85
USD
102896-414
102896-416
102896-418
(1R,2S,5S)-Rel-3-azabicyclo[3.1.0]hexane-2-methanol ≥95%
(1R,2S,5S)-Rel-3-azabicyclo[3.1.0]hexane-2-methanol
Hazard: Irritant
Formula:
C₆H₁₁NO |
MDL Number:
MFCD15072017 CAS Number: 1092506-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...