3-Azabicyclo[3.1.1]heptane ≥95%

Supplier: Matrix Scientific

115469-100MG 115469-250MG 115469-500MG
102896-378EA 1318.46 USD
102896-378 102896-410 102896-412
3-Azabicyclo[3.1.1]heptane ≥95%
3-Azabicyclo[3.1.1]heptane

Hazard: Irritant

Formula: C₆H₁₁N
MDL Number: MFCD19442693
CAS Number: 286-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR