N-Acetyl-3-hydroxy-L-phenylalanine ≥95%

Supplier: Matrix Scientific

115255-500MG 115255-1G
102895-740EA 598.46 USD
102895-740 102895-742
N-Acetyl-3-hydroxy-L-phenylalanine ≥95%
N-Acetyl-3-hydroxy-L-phenylalanine

Hazard: Irritant

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD01632511
CAS Number: 64325-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR