3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione ≥97%

Supplier: Matrix Scientific

115041-500MG 115041-1G
102895-258EA 693.85 USD
102895-258 102895-260
3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione ≥97%
3-(3-Methylphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione

Hazard: Irritant

Formula: C₁₅H₁₈N₂S
MDL Number: MFCD22549244
CAS Number: 5955-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR