6-Fluoro-4-methoxy-8-methylquinoline-2-carboxylic acid ≥90%

Supplier: Matrix Scientific

115040-500MG 115040-1G
102895-254EA 693.85 USD
102895-254 102895-256
6-Fluoro-4-methoxy-8-methylquinoline-2-carboxylic acid ≥90%
6-Fluoro-4-methoxy-8-methylquinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀FNO₃
MDL Number: MFCD21086559
CAS Number: 1338661-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR