Benzyl-3-hydroxy-3-methyl-azetidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115311-250MG 115311-500MG 115311-100MG
102895-042EA 1292.31 USD
102895-042 102895-044 102895-994
Benzyl-3-hydroxy-3-methyl-azetidine-1-carboxylate ≥95%
Benzyl-3-hydroxy-3-methyl-azetidine-1-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
MW: 221.26 g/mol
MDL Number: MFCD20528710
CAS Number: 1257294-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR