1-(Oxetan-3-yl)piperazine oxalate

Supplier: Matrix Scientific

113364-1G 113364-5G
102891-984EA 436.92 USD
102891-984 102891-986
1-(Oxetan-3-yl)piperazine oxalate
1-(Oxetan-3-yl)piperazine oxalate

Hazard: Irritant

Formula: C₉H₁₆N₂O₅
MDL Number: MFCD26959167
CAS Number: 1706458-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR