1-(Oxetan-3-yl)piperazine

Supplier: Matrix Scientific

113363-1G 113363-5G
102891-980EA 223.08 USD
102891-980 102891-982
1-(Oxetan-3-yl)piperazine
1-(Oxetan-3-yl)piperazine

Hazard: Irritant

Formula: C₇H₁₄N₂O
MDL Number: MFCD21847918
CAS Number: 1254115-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR