Anti-CD13 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 22A5]

Supplier: Southern Biotechnology
9556-02 9556-02S
101442-954EA 265.46 USD
101442-954 103265-316
Anti-CD13 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 22A5]
Antibodies
CD13, also known as aminopeptidase N, is a 150 kDa type II integral membrane glycoprotein. It is expressed on granulocytes, monocytes and their precursors, and on mast cells. CD13 is a zinc-binding metalloprotease that is involved in cell surface antigen presentation via its digestion of many MHC-bound regulatory peptides.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: 22A5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR