Anti-CD23 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: Tü1]

Supplier: Southern Biotechnology
9580-09 9580-09S
100246-206EA 329.54 USD
100246-206 103265-364
Anti-CD23 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: Tü1]
Antibodies
CD23 is a 45 kDa type II integral membrane glycoprotein that is the low affinity receptor for IgE. It also binds to CD21 and the α chains of CD11b and CD11c. It is expressed by B cells and monocytes and, weakly, on T cells, follicular dendritic cells, eosinophils, NK cells, Langerhans cells, and platelets. It is upregulated in response to B cell activation, CD40 ligation, or in response to IL-4 or IL-13. CD23 is responsible for regulation of IgE synthesis and may play a role in antigen presentation by B cells and follicular dendritic cells by interacting with CD40.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: Tü1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR