SpectraDye Antibody Labeling Kits, Advansta
Supplier: ADVANSTA INC MS
Make your own fluorescent antibody in one easy step.
- Quick and easy labeling in 30 minutes
- Labeled antibodies are compatible with IF microscopy, flow cytometry, Western blotting
- Choose from 6 common fluorescent dyes
- Once your primary antibody is labeled, no secondary is needed
- Labeling reaction requires as little as 10 micrograms antibody
SpectraDye Antibody Labeling Kits include sufficient reagents to label up to 1 mg of antibody
The SpectraDye antibody labeling kit allows for directly labeling primary antibodies, which means lower background in immunofluorescence-based experiments, reproducible single-color fluorescent applications, and flexibility for multicolor fluorescent applications.
SpectraDye dye kits contain Antibody Labeling Buffer, SpectraDye Dye Solution, Quenching Solution, and Neutralization Buffer. The user-supplied dye kit does not contain dye, but does contain high-quality DMSO to prepare your own dye solution.
Información para pedidos: Kit includes labeling buffer, SpectraDye dye solution (except user's dye kit), quenching solution, and neutralization buffer.