WesternBright MCF Western Blotting Kit, Advansta
Supplier: ADVANSTA INC MS
K-12022-010 K-12023-010
103254-712EA 269.79 USD
103254-712 103254-714 103254-716
WesternBright MCF Western Blotting Kit, Advansta
Blotting Kits
Multi-color fluorescent Western blotting in one convenient kit.
  • Multiplex - visualize two proteins at once
  • With pg detection, 10x more sensitive than Cy dyes
  • Only 3.5 hrs for entire protocol
  • Includes quality Western blotting reagents from Advansta
  • Compatible with Cy3/5 detecting or near-IR systems
  • Choose between visible (APC/RPE) or near-IR (IR700/800) kits
WesternBright MCF visible and near infrared (IR) fluorescent Western blotting kits allow the assay of two proteins at once, increasing the quality and quantity of information that can be gained from a single blot.
Información para pedidos: Kit includes secondary antibodies, blocking solution, washing solution, pre-cut PVDF membranes, background quenching sheets.