WesternBright Quantum HRP Substrate, Advansta
Supplier: ADVANSTA INC MS
K-12042-D10 K-12042-D20
103254-878EA 353.08 USD
103254-878 103254-880
WesternBright Quantum HRP Substrate, Advansta
Electrophoresis Reagents Western Blotting Reagents
Advansta's Quantum is ideal for quantitative chemiluminescent Western blotting.
  • Sensitive enough for attomole detection
  • Quantitative, with the linear range >3 orders magnitude
  • Optimized for CCD imaging and compatible with film
  • Low background and high signal to noise ratio
  • Long lasting signal, hours after substrate added
WesternBright Quantum chemiluminescent substrate sets the bar for sensitivity and quantitative ability. Specially developed for CCD imaging, WesternBright Quantum produces a strong, long-lasting signal with extremely low background, perfect for detecting low abundance proteins. Since it does not exhibit substrate depletion at high protein loads, WesternBright Quantum provides the largest dynamic range of any chemiluminescent substrate for the most quantitative chemiluminescent Western experiments.
Información para pedidos: Includes WesternBright Quantum luminol/enhancer solution and WesternBright Quantum stabilized peroxide solution.