TAE Buffers, concentrate, Molecular Biology Grade (Tris-acetate-EDTA), Promega
Supplier: Promega Corporation
V4271
V4281
PAV4271EA
82.51
USD
PAV4271
PAV4281
TAE Buffers, concentrate, Molecular Biology Grade (Tris-acetate-EDTA), Promega
Buffers
Electrophoresis Buffers
Running Buffers
TAE Buffer is the most commonly used buffer for agarose DNA electrophoresis.
- Quality tested and certified free of DNase, RNase and protease activity
- Composition (10X): 400 mM Tris-acetate, 10 mM EDTA
- Composition (40X): 1.6 M Tris-acetate, 40 mM EDTA
- Lead: ≤10ppm
- pH at 25°C: 8.2 - 8.4
A 1X solution is obtained by adding one part of the concentrated TAE to nine or 39 parts of deionized water.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...