D-(-)-Luciferin potassium salt

Supplier: Promega Corporation

Synonyms: (S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, Beetle Luciferin potassium salt, D-Luciferin potassium, Luciferin potassium salt, Firefly luciferin potassium salt, Potassium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate

E1602 E1603 E1601 E1605
PAE1602EA 254.04 USD
PAE1602 PAE1603 PAE1601 PAE1605
D-(-)-Luciferin potassium salt
D(-)-Luciferin potassium salt

Beetle luciferin (also known as D-luciferin) is synthesized as the monopotassium salt and is a substrate for the beetle luciferase reporter systems.


  • Use in solution assays in which the production of light can be monitored with either a luminometer or a scintillation counter
  • Can penetrate cell membranes, allowing transformed cells to be monitored for luciferase activity


Luciferase genes from the North American firefly (Photinus pyralis) and from other beetles are commonly used as reporter genes for studying transcription regulation in transient assay systems and as markers for stably transformed eukaryotic cells. Beetle luciferin (also known as D-luciferin) is synthesized as the monopotassium salt and is a substrate for the beetle luciferase reporter systems. D-luciferin is provided for those researchers who prefer to formulate their own assay reagents for monitoring in vitro or in vivo luciferase activity.

Formula: C₁₁H₇KN₂O₃S₂
MW: 318.41 g/mol
MDL Number: MFCD00044928
CAS Number: 115144-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR