Cetrimonium bromide ∼98% for molecular biology

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

0219400480 0219400483 0219400490 0219400491
IC0219400480EA 100.8 USD
IC0219400480 IC0219400483 IC0219400490 IC0219400491
Cetrimonium bromide ∼98% for molecular biology
Cetrimonium bromide

Cetyltrimethylammonium bromide is a phase transfer catalyst ideal for the precipitation of DNA. Hexadecyltrimethylammonium bromide is a bactericidal, cationic detergent. Its activity is neutralized by soaps and anionic detergents (i.e., SDS). Trimethylammonium bromide compounds form insoluble complexes with SDS. It is active at alkaline pH against both Gram positive and Gram negative organisms.


  • Storage temperature: Room Temperature (15 to 30 °C)


Cetyltrimethylammonium bromide ia used as an cationic surfactant suitable for the isolation of high molecular weight DNA in plants and other organisms (by a rapid method) as well as plant DNA for use in PCR analysis. It has been used to precipitate nucleic acids. CTAB provides enhancement of Concanavalin A mediated agglutination. It has been used for the determination of protein molecular weights in electrophoretic systems and for determination of critical micelle concentration of detergents. CTAB has also been used as a titrant for potentiometric titration of perchlorate and as a phase-transfer catalyst in reduction of arenes and heterocyclic compounds.

Formula: CH₃(CH₃)₁₅N(Br)(CH₃)₃
MW: 364.45 g/mol
Melting Pt: 248…251 °C (lit.)
Flash Pt: 244 °C (lit.)
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9 III
Merck Index: 13,02034

Order Now

Specification Test Results

Physical Appearance Powder
pH 6.0 - 7.5 (0.1 M H2O, 0 °C)(Lit.)
Aggregation Number 170
Critical Micelle Concentration 1 mM (20-25°C)
Solubility Freely but slowly soluble in water; Freely soluble in methanol and ethanol; soluble in alcohol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR