ONE-Glo™ Luciferase Assay System, Promega
Supplier: Promega Corporation
The ONE-Glo Luciferase Assay System provides a sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideal for high- and ultrahigh-throughput applications.
- Robust Performance for High- and Ultrahigh-Throughput Reporter Assays
- Less sensitive to mixing and dispensing conditions, enhancing reproducibility
- Ideal for use in high-density (384- and 1536-well) microplates
- Reduced odor, and convenient storage options
The ONE-Glo Luciferase Assay System provides a highly sensitive, robust, homogeneous assay for detection of firefly luciferase reporter gene expression in mammalian cells. Ideally suited for high- and ultrahigh-throughput applications, the ONE-Glo Assay contains a new luciferase substrate, resulting in a reagent that is more stable, more tolerant to sample components, and has less odor than standard luciferase assay reagents. These features ensure that the ONE-Glo Assay provides robust performance and also eliminates many of the handling inconveniences experienced using other reporter assays in a high-throughput setting.
These features ensure that the ONE-Glo Assay provides robust performance and also eliminates many of the handling inconveniences experienced using other reporter assays in a high-throughput setting.