Sunflower seed oil

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Helianthus Annuus (Sunflower) Seed Oil

S1929-200LTB S1929-20LT
700011-926EA 1655.25 USD
700011-926 700011-928
Sunflower seed oil
Sunflower seed oil

Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines

Density: 0.90
Storage Temperature: Ambient
MDL Number: MFCD00132403
CAS Number: 8001-21-6

Order Now

Specification Test Results

Synonyms Helianthus Annuus (Sunflower) Seed Oil
Free Fatty Acids APPROX. 0.13 %
Iodine Value Min. 110 - 143
Saponification Value Min. 188 - 194
Peroxide Value Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR