AlphaTec® 58-430 Nitrile Gloves with Grip Coating, Ansell
Supplier: Ansell Healthcare
256243 256244 256245 256246 256247
10769-504CS 1026.22 USD
10769-504 10769-506 10769-508 10769-510 10769-512
AlphaTec® 58-430 Nitrile Gloves with Grip Coating, Ansell
Gloves
These gloves feature Ansell GRIP™ Technology, which creates safe and secure glove contact with tools or part surfaces by wicking liquids into glove coating micro-channels
  • Cotton flocked lining helps to manage perspiration
  • Foldable gutter cuff helps to prevent dripping onto the forearm
  • Superior snag, puncture, and abrasion protection
The AlphaTec® 58-430 is a medium-duty glove with a 10 mil shell (2 mil grip) to protect against exposure to bases, oils, fuels, some solvents, greases, and animal fats. Snug, second-skin tailoring tightens the fit across the palm for added comfort and tactility and improves dexterity when moving between tasks.
This ensures optimal control and confidence when handling oily or wet objects.